Assay Design
Gene Transcripts
- Gene Symbol
- Ccdc77
- Entrez Gene ID
-
67200
- Gene Name
- coiled-coil domain containing 77
- Gene Aliases
-
2400002C23Rik, 2700091N06Rik, AV168274
- Chromosome Location
-
Chr.6: 120324322 - 120364369 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_026028.5 |
NP_080304.1 |
2 - 3 |
191 |
81 |
GenBank mRNA |
AK012598.1 |
- |
2 - 3 |
83 |
81 |
View Full Product Details