Assay Design



Gene Transcripts

Gene Symbol
Dph7
Entrez Gene ID
67228
Gene Name
diphthamine biosynethesis 7
Gene Aliases
2810443J12Rik, AW060693, Wdr85
Chromosome Location
Chr.2: 24962171 - 24973471 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026044.3 NP_080320.1 1 - 2 205 77
GenBank mRNA AK013297.1 - 1 - 2 185 77
AK078448.1 - 1 - 2 168 77
AK087998.1 - 1 - 2 168 77
AK152000.1 - 1 - 2 206 77
AK153537.1 - 1 - 2 206 77
BC086486.1 - 1 - 2 204 77

View Full Product Details