Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026464.2 | NP_080740.2 | 5 - 6 | 716 | 79 |
GenBank mRNA | AK018469.1 | - | 5 - 6 | 697 | 79 |
AK075958.1 | - | 5 - 6 | 716 | 79 | |
AK146093.1 | - | 5 - 6 | 735 | 79 | |
BC011484.1 | - | 3 - 4 | 382 | 79 | |
BC064750.1 | - | 5 - 6 | 675 | 79 | |
BC131932.1 | - | 5 - 6 | 710 | 79 | |
BC131934.1 | - | 5 - 6 | 710 | 79 |