Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027222.3 | NP_081498.2 | 1 - 2 | 202 | 109 |
GenBank mRNA | AK008016.1 | - | 1 - 2 | 203 | 109 |
AK088094.1 | - | 1 - 2 | 222 | 109 | |
BC064044.1 | - | 1 - 2 | 189 | 109 | |
GQ477351.1 | - | 1 - 2 | 168 | 109 |