Assay Design



Gene Transcripts

Gene Symbol
Slc25a33
Entrez Gene ID
70556
Gene Name
solute carrier family 25, member 33
Gene Aliases
5730438N18Rik, Pnc1
Chromosome Location
Chr.4: 149744036 - 149774267 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027460.2 NP_081736.2 6 - 7 976 71
GenBank mRNA AK017628.1 - 6 - 7 888 70
AK157423.1 - 6 - 7 977 71
AW989568.1 - 2 - 3 317 71
BC011293.1 - 6 - 7 951 71

View Full Product Details