Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001128180.1 | NP_001121652.1 | 8 - 9 | 1766 | 87 |
NM_001128181.1 | NP_001121653.1 | 9 - 10 | 1768 | 87 | |
NM_027712.3 | NP_081988.3 | 9 - 10 | 2753 | 87 | |
NM_177639.6 | NP_808307.2 | 10 - 11 | 2783 | 87 | |
GenBank mRNA | AK016873.1 | - | 1 - 2 | 209 | 87 |
AK034182.1 | - | 4 - 5 | 642 | 87 | |
AK035763.1 | - | 13 - 14 | 3334 | 87 | |
AK136926.1 | - | 9 - 10 | 1729 | 87 | |
AK147448.1 | - | 13 - 14 | 3570 | 87 | |
AK161941.1 | - | 8 - 9 | 1741 | 87 | |
AK220515.1 | - | 12 - 13 | 3633 | 87 | |
AY243846.2 | - | 9 - 10 | 2534 | 87 | |
BC062120.1 | - | 10 - 11 | 2784 | 87 | |
BC094369.1 | - | 8 - 9 | 1962 | 87 |