Assay Design



Gene Transcripts

Gene Symbol
Tmem68
Entrez Gene ID
72098
Gene Name
transmembrane protein 68
Gene Aliases
2010300G19Rik, AA408325, AW123402
Chromosome Location
Chr.4: 3549041 - 3574848 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028097.3 NP_082373.1 7 - 8 1066 81
GenBank mRNA AK008493.1 - 7 - 8 1100 81
AK048210.1 - 7 - 8 1066 81
AK084881.1 - 7 - 8 1100 81
AK088600.1 - 8 - 9 1236 81
AK147765.1 - 7 - 8 1099 81
AK162563.1 - 7 - 8 1099 81
AK165916.1 - 7 - 8 1128 81
AK166237.1 - 7 - 8 1152 81
BC016240.1 - 7 - 8 1084 81

View Full Product Details