Assay Design



Gene Transcripts

Gene Symbol
Zdhhc4
Entrez Gene ID
72881
Gene Name
zinc finger, DHHC domain containing 4
Gene Aliases
1810021D01Rik, 2900029I10Rik
Chromosome Location
Chr.5: 143316489 - 143329252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028379.5 NP_082655.1 6 - 7 756 73
GenBank mRNA AK007572.1 - 4 - 5 742 73
AK013609.1 - 6 - 7 743 73
BC019523.1 - 6 - 7 738 73

View Full Product Details