Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028715.3 | NP_082991.3 | 22 - 23 | 2002 | 77 |
NR_028267.1 | 22 - 23 | 2002 | 77 | ||
GenBank mRNA | AK014382.1 | - | 22 - 23 | 1987 | 77 |
AK137333.1 | - | 22 - 23 | 1990 | 77 | |
AK172927.1 | - | 14 - 15 | 3049 | 77 | |
BC032207.1 | - | 21 - 22 | 1978 | 77 |