Assay Design



Gene Transcripts

Gene Symbol
Acoxl
Entrez Gene ID
74121
Gene Name
acyl-Coenzyme A oxidase-like
Gene Aliases
1200014P05Rik, AV025673
Chromosome Location
Chr.2: 127838797 - 128126252 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028765.3 NP_083041.1 12 - 13 1434 132
GenBank mRNA AK004775.1 - 12 - 13 1436 132
BC048717.1 - 5 - 6 530 132

View Full Product Details