Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166506.1 | NP_001159978.1 | 10 - 11 | 1380 | 70 |
NM_001166507.1 | NP_001159979.1 | 10 - 11 | 1380 | 70 | |
NM_028777.3 | NP_083053.2 | 10 - 11 | 1380 | 70 | |
GenBank mRNA | AK004818.1 | - | 10 - 11 | 1305 | 70 |
AK047396.1 | - | 10 - 11 | 1388 | 70 | |
AK172947.1 | - | 7 - 8 | 915 | 70 | |
AK193303.1 | - | 1 - 2 | 59 | 70 | |
BC005766.1 | - | 10 - 11 | 1338 | 70 | |
BC086620.1 | - | 10 - 11 | 1314 | 70 |