Assay Design



Gene Transcripts

Gene Symbol
Slc35f5
Entrez Gene ID
74150
Gene Name
solute carrier family 35, member F5
Gene Aliases
1300003P13Rik, AI646727
Chromosome Location
Chr.1: 125560414 - 125595780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028787.4 NP_083063.2 1 - 2 273 66
GenBank mRNA AK004892.2 - 1 - 2 254 66
AK076339.1 - 1 - 2 267 66
AK145848.1 - 1 - 2 274 66
AK169012.1 - 1 - 2 135 66
AK171951.1 - 1 - 2 247 66
BC026858.1 - 1 - 2 132 66

View Full Product Details