Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028841.3 | NP_083117.2 | 7 - 8 | 936 | 89 |
GenBank mRNA | AK008761.1 | - | 7 - 8 | 881 | 89 |
AK166272.1 | - | 7 - 8 | 939 | 89 | |
AK171691.1 | - | 7 - 8 | 885 | 89 | |
BC010346.1 | - | 7 - 8 | 881 | 89 | |
CD740983.1 | - | 2 - 3 | 89 |