Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033654.2 | NP_001028826.1 | 5 - 6 | 573 | 63 |
NM_028304.2 | NP_082580.1 | 5 - 6 | 986 | 63 | |
NM_028956.4 | NP_083232.1 | 5 - 6 | 583 | 63 | |
GenBank mRNA | AK012737.1 | - | 5 - 6 | 750 | 63 |
AK017063.1 | - | 5 - 6 | 560 | 63 | |
AK028368.1 | - | 5 - 6 | 570 | 63 | |
AK029866.1 | - | 5 - 6 | 574 | 63 | |
AK088562.1 | - | 5 - 6 | 574 | 63 | |
AK145055.1 | - | 5 - 6 | 932 | 63 | |
BC025555.1 | - | 5 - 6 | 804 | 63 |