Assay Design



Gene Transcripts

Gene Symbol
Rprd2
Entrez Gene ID
75137
Gene Name
regulation of nuclear pre-mRNA domain containing 2
Gene Aliases
2810036A19Rik, 4930535B03Rik, 6720469I21Rik, AL022841, AL022940, AU021304, BB077382, mKIAA0460
Chromosome Location
Chr.3: 95759873 - 95819723 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081293.1 NP_001074762.1 4 - 5 771 97
GenBank mRNA AK015972.1 - 4 - 5 628 97
AK083674.1 - 4 - 5 540 97
AK087157.1 - 4 - 5 543 97
BC067054.1 - 4 - 5 576 97

View Full Product Details