Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029466.4 | NP_083742.2 | 5 - 6 | 658 | 87 |
GenBank mRNA | AK016359.1 | - | 5 - 6 | 615 | 87 |
AK040422.1 | - | 5 - 6 | 701 | 87 | |
AK041237.1 | - | 5 - 6 | 689 | 87 | |
AK042200.1 | - | 5 - 6 | 744 | 87 | |
AK045310.1 | - | - | 736 | 87 | |
AK053731.1 | - | 5 - 6 | 659 | 87 | |
AK164341.1 | - | 5 - 6 | 653 | 87 | |
BC066810.1 | - | 5 - 6 | 697 | 87 |