Assay Design



Gene Transcripts

Gene Symbol
Nadsyn1
Entrez Gene ID
78914
Gene Name
NAD synthetase 1
Gene Aliases
9130012B15Rik
Chromosome Location
Chr.7: 143795587 - 143822864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ305343.1 - 8 - 9 766 71
AK020264.1 - 8 - 9 766 71
AK041101.1 - 8 - 9 753 71
AK042389.1 - 8 - 9 726 71
AK045774.1 - 8 - 9 753 71
AK046491.1 - 8 - 9 753 71
BC038016.1 - 8 - 9 712 71

View Full Product Details