Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133937.1 | NP_598698.1 | 2 - 3 | 210 | 70 |
GenBank mRNA | AK003342.1 | - | 2 - 3 | 225 | 70 |
AK020128.1 | - | 2 - 3 | 225 | 70 | |
AK020277.1 | - | 2 - 3 | 226 | 70 | |
AK150689.1 | - | 2 - 3 | 225 | 70 | |
AK163236.1 | - | 2 - 3 | 225 | 70 | |
AK190298.1 | - | 2 - 3 | 149 | 70 | |
AK193342.1 | - | 2 - 3 | 149 | 70 | |
BC010582.1 | - | 2 - 3 | 210 | 70 |