Assay Design



Gene Transcripts

Gene Symbol
Ttc27
Entrez Gene ID
74196
Gene Name
tetratricopeptide repeat domain 27
Gene Aliases
2610511O17Rik
Chromosome Location
Chr.17: 74717736 - 74863570 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152817.4 NP_690030.3 2 - 3 445 70
GenBank mRNA AK012131.1 - 2 - 3 445 70
AK030942.1 - 2 - 3 446 70
AK049324.1 - 2 - 3 446 70
AK168138.1 - 2 - 3 436 70
BC021912.1 - 1 - 2 180 70

View Full Product Details