Assay Design



Gene Transcripts

Gene Symbol
Cntn1
Entrez Gene ID
12805
Gene Name
contactin 1
Gene Aliases
AW495098, CNTN, F3cam
Chromosome Location
Chr.15: 92051142 - 92341967 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159647.1 NP_001153119.1 19 - 20 2803 71
NM_001159648.1 NP_001153120.1 19 - 20 2657 71
NM_007727.2 NP_031753.1 19 - 20 2706 71
GenBank mRNA AK076273.1 - 5 - 6 710 71
BC066864.1 - 19 - 20 2729 71
X14943.1 - 19 - 20 2676 71

View Full Product Details