Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252653.1 | NP_001239582.1 | 6 - 7 | 569 | 57 |
NM_007899.2 | NP_031925.2 | 6 - 7 | 572 | 57 | |
GenBank mRNA | AK153009.1 | - | 6 - 7 | 571 | 57 |
AK153471.1 | - | 6 - 7 | 571 | 57 | |
AK159334.1 | - | 6 - 7 | 568 | 57 | |
AK169939.1 | - | 6 - 7 | 574 | 57 | |
BC138693.1 | - | 6 - 7 | 558 | 57 | |
BC145381.1 | - | 6 - 7 | 555 | 57 | |
L33416.1 | - | 6 - 7 | 565 | 57 |