Assay Design



Gene Transcripts

Gene Symbol
Eps15
Entrez Gene ID
13858
Gene Name
epidermal growth factor receptor pathway substrate 15
Gene Aliases
2410112D09Rik
Chromosome Location
Chr.4: 109280256 - 109387817 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159964.1 NP_001153436.1 6 - 7 1074 114
NM_007943.3 NP_031969.1 18 - 19 2023 114
GenBank mRNA AK083176.1 - 6 - 7 1058 114
AY190610.1 - 18 - 19 1996 114
BC048783.1 - 18 - 19 1997 114
L21768.1 - 18 - 19 1996 114

View Full Product Details