Assay Design
Gene Transcripts
- Gene Symbol
- Fmo3
- Entrez Gene ID
-
14262
- Gene Name
- flavin containing monooxygenase 3
- Gene Aliases
-
AW111792
- Chromosome Location
-
Chr.1: 162953800 - 162984491 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_008030.1 |
NP_032056.1 |
1 - 2 |
35 |
61 |
GenBank mRNA |
U87147.1 |
- |
1 - 2 |
35 |
61 |
View Full Product Details