Assay Design



Gene Transcripts

Gene Symbol
Brca1
Entrez Gene ID
12189
Gene Name
breast cancer 1
Gene Aliases
-
Chromosome Location
Chr.11: 101488754 - 101551956 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009764.3 NP_033894.3 13 - 14 4561 67
GenBank mRNA AB221610.1 - 4 - 5 1790 67
BC068303.1 - 13 - 14 4397 67
U31625.1 - 13 - 14 4418 67
U32446.1 - 13 - 14 4471 67
U35641.1 - 13 - 14 4454 67
U36475.1 - 12 - 13 4381 67
U68174.1 - 13 - 14 4465 67

View Full Product Details