Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001109764.1 | NP_001103234.1 | 7 - 8 | 1198 | 98 |
NM_009819.2 | NP_033949.2 | 6 - 7 | 1096 | 98 | |
NM_145732.2 | NP_663785.2 | 7 - 8 | 1198 | 98 | |
GenBank mRNA | AK035033.1 | - | 6 - 7 | 992 | 98 |
AK158916.1 | - | 7 - 8 | 1231 | 98 | |
BC051995.1 | - | 6 - 7 | 977 | 98 | |
BC079648.1 | - | 7 - 8 | 1115 | 98 | |
D25281.1 | - | 7 - 8 | 1125 | 98 | |
D25282.1 | - | 6 - 7 | 1025 | 98 |