Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043317.2 | NP_001036782.1 | 5 - 6 | 802 | 75 |
NM_009845.3 | NP_033975.3 | 3 - 4 | 677 | 75 | |
NR_102722.1 | 5 - 6 | 876 | 75 | ||
NR_102723.1 | 5 - 6 | 816 | 75 | ||
GenBank mRNA | AF102134.1 | - | 2 - 3 | 446 | 75 |
AF115400.1 | - | 2 - 3 | 446 | 75 | |
AF115401.1 | - | 2 - 3 | 446 | 75 | |
AJ250676.1 | - | 2 - 3 | 589 | 75 | |
AJ250677.1 | - | 2 - 3 | 576 | 75 | |
AJ250678.1 | - | 2 - 3 | 558 | 75 | |
AJ250679.1 | - | 2 - 3 | 521 | 75 | |
AJ250680.1 | - | 3 - 4 | 490 | 75 | |
AJ250681.1 | - | - | 421 | 75 | |
AJ250683.1 | - | - | 372 | 75 | |
AJ250684.1 | - | 3 - 4 | 335 | 75 | |
AK041230.1 | - | 5 - 6 | 811 | 75 | |
AK143835.1 | - | 5 - 6 | 740 | 75 | |
AK156735.1 | - | 3 - 4 | 676 | 75 | |
AK171589.1 | - | 2 - 3 | 483 | 75 | |
AK171990.1 | - | 2 - 3 | 797 | 75 | |
AK172301.1 | - | 3 - 4 | 633 | 75 | |
BC046957.1 | - | 5 - 6 | 728 | 75 | |
BC150807.1 | - | 4 - 5 | 761 | 75 | |
L02844.1 | - | 4 - 5 | 948 | 75 | |
L16928.1 | - | 4 - 5 | 925 | 75 |