Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001043317.2 | NP_001036782.1 | 11 - 12 | 2465 | 88 |
NM_009845.3 | NP_033975.3 | 9 - 10 | 2340 | 88 | |
NR_102722.1 | 11 - 12 | 2537 | 88 | ||
NR_102723.1 | 11 - 12 | 2498 | 88 | ||
GenBank mRNA | AF102134.1 | - | 8 - 9 | 2109 | 88 |
AF115400.1 | - | 8 - 9 | 2109 | 88 | |
AF115401.1 | - | 8 - 9 | 2109 | 88 | |
AK041230.1 | - | 11 - 12 | 2472 | 88 | |
AK143835.1 | - | 11 - 12 | 2403 | 88 | |
AK171589.1 | - | 8 - 9 | 2146 | 88 | |
BC046957.1 | - | 11 - 12 | 2410 | 88 | |
L16928.1 | - | 10 - 11 | 2588 | 88 |