Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170744.1 | NP_001164215.1 | 4 - 5 | 2241 | 61 |
NM_009980.4 | NP_034110.1 | 6 - 7 | 1118 | 61 | |
GenBank mRNA | AB033123.1 | - | 6 - 7 | 983 | 61 |
AF016508.1 | - | 4 - 5 | 590 | 61 | |
AF059735.1 | - | 4 - 5 | 569 | 61 | |
AK134399.1 | - | 5 - 6 | 642 | 61 | |
AK146713.1 | - | 6 - 7 | 1003 | 61 | |
AK147871.1 | - | 6 - 7 | 1001 | 61 | |
BC013333.1 | - | 4 - 5 | 625 | 61 | |
BC017520.1 | - | 5 - 6 | 2086 | 61 |