Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009984.3 | NP_034114.1 | 7 - 8 | 983 | 87 |
GenBank mRNA | AF121837.1 | - | 7 - 8 | 947 | 87 |
AF121838.1 | - | 7 - 8 | 947 | 87 | |
AF121839.1 | - | 7 - 8 | 947 | 87 | |
AK002221.1 | - | 7 - 8 | 983 | 87 | |
AK011589.1 | - | 7 - 8 | 983 | 87 | |
AK049466.1 | - | 7 - 8 | 984 | 87 | |
AK136341.1 | - | 6 - 7 | 824 | 87 | |
AK145051.1 | - | 7 - 8 | 984 | 87 | |
AK147142.1 | - | 7 - 8 | 985 | 87 | |
AK150452.1 | - | 7 - 8 | 984 | 87 | |
AK151169.1 | - | 7 - 8 | 984 | 87 | |
AK153076.1 | - | 7 - 8 | 984 | 87 | |
AK153417.1 | - | 7 - 8 | 984 | 87 | |
AK159511.1 | - | 7 - 8 | 984 | 87 | |
AK160100.1 | - | 7 - 8 | 984 | 87 | |
AK161538.1 | - | 7 - 8 | 984 | 87 | |
AK165397.1 | - | 7 - 8 | 984 | 87 | |
AK169394.1 | - | 7 - 8 | 984 | 87 | |
AK169712.1 | - | 7 - 8 | 984 | 87 | |
BC068163.1 | - | 7 - 8 | 969 | 87 | |
J02583.1 | - | 7 - 8 | 960 | 87 | |
X06086.1 | - | 7 - 8 | 969 | 87 |