Assay Design



Gene Transcripts

Gene Symbol
Ctsw
Entrez Gene ID
13041
Gene Name
cathepsin W
Gene Aliases
lymphopain
Chromosome Location
Chr.19: 5465240 - 5470387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009985.4 NP_034115.2 6 - 7 627 94
GenBank mRNA AF014941.1 - 6 - 7 612 94
AK088375.1 - 6 - 7 628 94
BC019136.1 - 5 - 6 705 94

View Full Product Details