Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010442.2 | NP_034572.1 | 1 - 2 | 159 | 72 |
GenBank mRNA | AK150934.1 | - | 1 - 2 | 156 | 72 |
AK151005.1 | - | 1 - 2 | 156 | 72 | |
AK151012.1 | - | 1 - 2 | 147 | 72 | |
AK151627.1 | - | 1 - 2 | 134 | 72 | |
AK153420.1 | - | 1 - 2 | 156 | 72 | |
AK153516.1 | - | 1 - 2 | 156 | 72 | |
AK153566.1 | - | 1 - 2 | 156 | 72 | |
AK154598.1 | - | 1 - 2 | 157 | 72 | |
AK159521.1 | - | 1 - 2 | 156 | 72 | |
AK159870.1 | - | 1 - 2 | 160 | 72 | |
AK159959.1 | - | 1 - 2 | 160 | 72 | |
AK169227.1 | - | 1 - 2 | 156 | 72 | |
BC010757.1 | - | 1 - 2 | 128 | 72 | |
BY319630.1 | - | 1 - 2 | 161 | 72 | |
M33203.1 | - | 1 - 2 | 101 | 72 | |
X13356.1 | - | 1 - 2 | 101 | 72 |