Assay Design



Gene Transcripts

Gene Symbol
Gtpbp1
Entrez Gene ID
14904
Gene Name
GTP binding protein 1
Gene Aliases
AL022987, AW045683, C85212, GP-1, Gtpbp
Chromosome Location
Chr.15: 79690896 - 79721479 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013818.2 NP_038846.2 4 - 5 846 59
GenBank mRNA AK004612.1 - 4 - 5 847 59
AK147717.1 - 4 - 5 848 59
AK150068.1 - 4 - 5 853 59
AK150185.1 - 4 - 5 853 59
AK158342.1 - 4 - 5 856 59
AK170091.1 - 4 - 5 848 59
BC037129.1 - 2 - 3 388 59
BC046228.1 - 4 - 5 846 59
U87965.1 - 4 - 5 733 59

View Full Product Details