Assay Design



Gene Transcripts

Gene Symbol
Fbxo18
Entrez Gene ID
50755
Gene Name
F-box protein 18
Gene Aliases
AU015756, Fbh1, Fbx18
Chromosome Location
Chr.2: 11742573 - 11777585 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_015792.1 NP_056607.1 17 - 18 2687 85
GenBank mRNA AF184275.1 - 15 - 16 2181 85
AK003964.1 - 2 - 3 248 85
AK147829.1 - 17 - 18 2671 85
AK154357.1 - 17 - 18 2710 85
AK158937.1 - 17 - 18 2738 85
AK219751.1 - 1 - 2 83 85
AK220003.1 - 1 - 2 83 85
BC004036.1 - 14 - 15 1773 85
BC031393.1 - 17 - 18 2687 85

View Full Product Details