Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018772.4 | NP_061242.3 | 1 - 2 | 294 | 137 |
GenBank mRNA | AF106967.1 | - | 1 - 2 | 213 | 137 |
AK049027.1 | - | 1 - 2 | 295 | 137 | |
BC005677.1 | - | 1 - 2 | 272 | 137 | |
BC021149.1 | - | 1 - 2 | 240 | 137 | |
BC095955.1 | - | 1 - 2 | 249 | 137 |