Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018788.3 | NP_061258.2 | 6 - 7 | 3210 | 59 |
GenBank mRNA | AF083550.1 | - | 4 - 5 | 3000 | 59 |
AK137842.1 | - | 4 - 5 | 2287 | 59 | |
AK220344.1 | - | 4 - 5 | 2553 | 59 | |
BC058526.1 | - | 4 - 5 | 2601 | 59 | |
BC065073.1 | - | 4 - 5 | 2675 | 59 |