Assay Design
Gene Transcripts
- Gene Symbol
- Angel2
- Entrez Gene ID
-
52477
- Gene Name
- angel homolog 2 (Drosophila)
- Gene Aliases
-
2610307I21Rik, 5730410O10Rik, AI845692, D1Ertd396e, D1Ertd654e
- Chromosome Location
-
Chr.1: 190925108 - 190946962 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AB041602.1 |
- |
4 - 5 |
989 |
110 |
AK207828.1 |
- |
1 - 2 |
|
110 |
View Full Product Details