Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023173.2 | NP_075662.2 | 1 - 2 | 474 | 84 |
GenBank mRNA | AF268196.1 | - | 1 - 2 | 399 | 84 |
AF280810.1 | - | 1 - 2 | 341 | 84 | |
AF280811.1 | - | 1 - 2 | 341 | 84 | |
AK004488.1 | - | 1 - 2 | 356 | 84 | |
BC099453.1 | - | 1 - 2 | 474 | 84 |