Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024204.6 | NP_077166.4 | 1 - 2 | 292 | 69 |
GenBank mRNA | AK012869.1 | - | 1 - 2 | 305 | 69 |
AK017360.1 | - | 1 - 2 | 293 | 69 | |
AK037242.1 | - | 1 - 2 | 175 | 69 | |
BC003460.1 | - | 1 - 2 | 154 | 69 |