Assay Design



Gene Transcripts

Gene Symbol
Ciao1
Entrez Gene ID
26371
Gene Name
cytosolic iron-sulfur protein assembly 1
Gene Aliases
AW210570, Wdr39
Chromosome Location
Chr.2: 127240938 - 127247816 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025296.4 NP_079572.2 5 - 6 891 80
GenBank mRNA AK002314.1 - 5 - 6 836 80
AK028376.1 - 5 - 6 818 80
BC004089.1 - 5 - 6 812 80

View Full Product Details