Assay Design



Gene Transcripts

Gene Symbol
Bzw2
Entrez Gene ID
66912
Gene Name
basic leucine zipper and W2 domains 2
Gene Aliases
1110001I24Rik, Bdm2, HSPC028
Chromosome Location
Chr.12: 36091835 - 36156825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025840.3 NP_080116.2 7 - 8 816 75
GenBank mRNA AB244988.1 - 7 - 8 732 75
AK003229.1 - 7 - 8 811 75
AK011253.1 - 7 - 8 772 75
AK088108.1 - 7 - 8 749 75
AK132595.1 - 7 - 8 758 75
AK160544.1 - 7 - 8 789 75
AK162647.1 - 7 - 8 758 75
AK205942.1 - 2 - 3 171 75
BC013060.1 - 7 - 8 752 75

View Full Product Details