Assay Design
Gene Transcripts
- Gene Symbol
- Zkscan6
- Entrez Gene ID
-
52712
- Gene Name
- zinc finger with KRAB and SCAN domains 6
- Gene Aliases
-
1700128E15Rik, D11Ertd714e, KOX11, Zfp535, Znf18
- Chromosome Location
-
Chr.11: 65807175 - 65829239 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK007308.1 |
- |
5 - 6 |
956 |
64 |
BC043075.1 |
- |
6 - 7 |
987 |
64 |
View Full Product Details