Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026222.2 | NP_080498.1 | 19 - 20 | 2778 | 94 |
GenBank mRNA | AK007153.1 | - | 3 - 4 | 281 | 94 |
AK014839.1 | - | 19 - 20 | 2770 | 94 | |
AK030087.1 | - | 19 - 20 | 2745 | 94 | |
BC138917.1 | - | 19 - 20 | 2740 | 94 | |
BC138920.1 | - | 19 - 20 | 2740 | 94 |