Assay Design



Gene Transcripts

Gene Symbol
Inpp4a
Entrez Gene ID
269180
Gene Name
inositol polyphosphate-4-phosphatase, type I
Gene Aliases
107kDa, 9630012D15, D130048C09Rik, R74740
Chromosome Location
Chr.1: 37299838 - 37410740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290797.1 NP_001277726.1 12 - 13 1495 64
NM_001290798.1 NP_001277727.1 2 - 3 371 64
NM_001290799.1 NP_001277728.1 2 - 3 371 64
NM_030266.4 NP_084542.2 12 - 13 1495 64
NM_172971.3 NP_766559.2 2 - 3 371 64
GenBank mRNA AF317838.1 - 10 - 11 1048 64
AK035867.1 - 2 - 3 372 64
AK149414.1 - 12 - 13 1420 64
BC120541.1 - 2 - 3 367 64
BC144747.1 - 2 - 3 367 64
BC144748.1 - 2 - 3 367 64

View Full Product Details