Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054094.5 | NP_473435.1 | 8 - 9 | 1191 | 83 |
GenBank mRNA | AB059428.1 | - | 8 - 9 | 1195 | 83 |
AK149550.1 | - | 8 - 9 | 1201 | 83 | |
BC016414.1 | - | 8 - 9 | 1188 | 83 | |
BC022149.1 | - | 8 - 9 | 1169 | 83 |