Assay Design



Gene Transcripts

Gene Symbol
Reep6
Entrez Gene ID
70335
Gene Name
receptor accessory protein 6
Gene Aliases
0610011M24Rik, Dp1l1
Chromosome Location
Chr.10: 80330003 - 80336441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204931.1 NP_001191860.1 1 - 2 207 73
NM_139292.2 NP_647453.1 1 - 2 207 73
GenBank mRNA AB039933.1 - 1 - 2 182 73
AK002562.1 - 1 - 2 205 73
AY562234.1 - 1 - 2 121 73
BC029741.1 - 1 - 2 194 73

View Full Product Details