Assay Design
Gene Transcripts
- Gene Symbol
- Reep6
- Entrez Gene ID
-
70335
- Gene Name
- receptor accessory protein 6
- Gene Aliases
-
0610011M24Rik, Dp1l1
- Chromosome Location
-
Chr.10: 80330003 - 80336441 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_139292.2 |
NP_647453.1 |
4 - 5 |
605 |
155 |
GenBank mRNA |
AB039933.1 |
- |
4 - 5 |
580 |
155 |
View Full Product Details