Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144553.2 | NP_653136.2 | 5 - 6 | 878 | 91 |
GenBank mRNA | AB076696.1 | - | 5 - 6 | 878 | 91 |
AK036384.1 | - | 5 - 6 | 680 | 91 | |
AK048470.1 | - | 5 - 6 | 648 | 91 | |
AK085882.1 | - | 5 - 6 | 659 | 91 | |
AK129032.1 | - | 5 - 6 | 624 | 91 | |
AK135820.1 | - | 5 - 6 | 685 | 91 | |
AK147955.1 | - | 5 - 6 | 648 | 91 | |
BC027273.1 | - | 5 - 6 | 618 | 91 | |
BC058087.1 | - | 5 - 6 | 612 | 91 |