Assay Design



Gene Transcripts

Gene Symbol
Fam49b
Entrez Gene ID
223601
Gene Name
family with sequence similarity 49, member B
Gene Aliases
0910001A06Rik, AW122079
Chromosome Location
Chr.15: 63929094 - 64060452 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144846.5 NP_659095.1 4 - 5 499 57
GenBank mRNA AK028062.1 - 4 - 5 500 57
AK075613.1 - 5 - 6 444 57
BC011343.1 - 4 - 5 317 57

View Full Product Details