Assay Design



Gene Transcripts

Gene Symbol
Dus3l
Entrez Gene ID
224907
Gene Name
dihydrouridine synthase 3-like (S. cerevisiae)
Gene Aliases
AI662135, AW557805
Chromosome Location
Chr.17: 56764717 - 56770093 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144858.2 NP_659107.2 6 - 7 1279 58
GenBank mRNA AK157702.1 - 6 - 7 1220 58
AK164614.1 - 5 - 6 879 58
AK166183.1 - 6 - 7 1173 58
AY040840.1 - 6 - 7 1252 58
CX201215.1 - 2 - 3 282 58

View Full Product Details