Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145353.2 | NP_663328.2 | 3 - 4 | 553 | 110 |
GenBank mRNA | AF197931.1 | - | 2 - 3 | 313 | 110 |
AK011822.1 | - | 3 - 4 | 540 | 110 | |
AK042644.1 | - | 2 - 3 | 225 | 110 | |
AK149957.1 | - | 3 - 4 | 386 | 110 | |
BC011161.1 | - | 3 - 4 | 514 | 110 | |
BC019384.1 | - | 3 - 4 | 534 | 110 | |
BC100575.1 | - | 3 - 4 | 485 | 110 |